Learn More
R&D Systems™ Recombinant Human Coagulation Factor II/Thrombin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 1473-SE-010
Product Code. 18107190
  • £427.00 / 10µg
Explore more special offers
Description
Description
The Recombinant Human Coagulation Factor II/Thrombin Protein is derived from NS0. The Recombinant Human Coagulation Factor II/Thrombin Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in HEPES and NaCl.
4 kDa (light chain A), 31 kDa (heavy chain B), 14 kDa (C-terminal fragment)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
2147
Coagulation Factor II/Thrombin
Mouse myeloma cell line,NS0-derived human Coagulation Factor II/Thrombin protein Met1-Glu622 with a C-terminal 10-His tag The proenzyme was purified,activated and further purified.
Active form
Recombinant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Recombinant Human Coagulation Factor II/Thrombin Protein > 10ug; Carrier Free