missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Coagulation Factor VII Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 2338-SE-010
This item is not returnable.
View return policy
Description
The Recombinant Human Coagulation Factor VII Protein is derived from NS0. The Recombinant Human Coagulation Factor VII Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2μm filtered solution in Sodium Acetate and NaCl. | |
46 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
2155 | |
Coagulation Factor VII | |
Mouse myeloma cell line,NS0-derived human Coagulation Factor VII protein Ala39-Pro444,with a C-terminal 10-His tag | |
Mature form | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human Coagulation Factor VII Protein > 10ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction