missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL7 / Interleukin 7 Protein
A DNA sequence encoding the Homo sapiens(Human) IL7 / Interleukin 7 Protein, was expressed in the hosts and tags indicated.
£428.07 - £1240.00
Specifications
Accession Number | NP_000871.1 |
---|---|
Gene ID (Entrez) | 3574 |
Name | IL7 / Interleukin 7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079131
|
enQuireBio™
QP9979-IV-100UG |
100 μg |
£1240.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16099131
|
enQuireBio™
QP9979-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089131
|
enQuireBio™
QP9979-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000871.1 | |
IL7 / Interleukin 7 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3574 | |
Research Use Only | |
IL7 | |
Recombinant Protein | |
E. coli | |
DCDIEGKDGK QYESVLMVSI DQLLDSMKEI GSNCLNNEFN FFKRHICDAN KEGMFLFRAA RKLRQFLKMN STGDFDLHLL KVSEGTTILL NCTGQVKGRK PAALGEAQPT KSLEENKSLK EQKKLNDLCF LKRLLQEIKT CWNKILMGTK EH | |
Greater than 85% as determined by SDS-PAGE. |