missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NK-tumor recognition Protein
A DNA sequence encoding the Homo sapiens (Human) NK-tumor recognition protein, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_005376.2 |
---|---|
Gene ID (Entrez) | 4820 |
Name | NK-tumor recognition Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073200
|
enQuireBio™
QP6422-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093200
|
enQuireBio™
QP6422-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083200
|
enQuireBio™
QP6422-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003210
|
enQuireBio™
QP6422-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013210
|
enQuireBio™
QP6422-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023210
|
enQuireBio™
QP6422-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005376.2 | |
NK-tumor recognition Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4820 | |
Research Use Only | |
NKTR | |
Recombinant Protein | |
E. coli | |
HFDIEINREP VGRIMFQLFS DICPKTCKNF LCLCSGEKGL GKTTGKKLCY KGSTFHRVVK NFMIQGGDFS EGNGKGGESI YGGYFKDENF ILKHDRAFLL SMANRGKHTN GSQFFITTKP APHLDGVHVV FGLVISGFEV IEQIENLKTD AASRPYADVR VIDCGV | |
Greater than 90% as determined by SDS-PAGE. |