missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S-phase kinase-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) S-phase kinase-associated protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6689-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_008861.2 | |
S-phase kinase-associated protein 1 | |
Research Use Only | |
SKP1 | |
Recombinant Protein | |
E. coli | |
PSIKLQSSDG EIFEVDVEIA KQSVTIKTML EDLGMDDEGD DDPVPLPNVN AAILKKVIQW CTHHKDDPPP PEDDENKEKR TDDIPVWDQE FLKVDQGTLF ELILAANYLD IKGLLDVTCK TVANMIKGKT PEEIRKTFNI KNDFTEEEEA QVRKENQWCE EK | |
Greater than 90% as determined by SDS-PAGE. |
6500 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human S-phase kinase-associated protein 1 > 1mg
Spot an opportunity for improvement?Share a Content Correction