missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine racemase Protein
A DNA sequence encoding the Homo sapiens (Human) Serine racemase, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_068766.1 |
---|---|
Gene ID (Entrez) | 63826 |
Name | Serine racemase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16005430
|
enQuireBio™
QP6729-ec-10ug |
10 μg | |||||||
16045430
|
enQuireBio™
QP6729-ec-50ug |
50 μg | |||||||
16095420
|
enQuireBio™
QP6729-ec-100ug |
100 μg |
£560.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16025430
|
enQuireBio™
QP6729-ec-200ug |
200 μg | |||||||
16035430
|
enQuireBio™
QP6729-ec-500ug |
500 μg | |||||||
16015430
|
enQuireBio™
QP6729-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_068766.1 | |
Serine racemase Protein | |
Not determined. | |
Full Length | |
Human | |
Tris-based buffer, 50% glycerol |
63826 | |
Research Use Only | |
SRR | |
Recombinant Protein | |
MCAQYCISFA DVEKAHINIR DSIHLTPVLT SSILNQLTGR NLFFKCELFQ KTGSFKIRGA LNAVRSLVPD ALERKPKAVV THSSGNHGQA LTYAAKLEGI PAYIVVPQTA PDCKKLAIQA YGASIVYCEP SDESRENVAK RVTEETEGIM VHPNQEPAVI AGQGTIALEV LNQVPLVDAL VVPVGGGGML AGIAITVKAL KPSVKVYAAE PSNADDCYQS KLKGKLMPNL YPPETIADGV KSSIGLNTWP IIRDLVDDIF TVTEDEIKCA TQLVWERMKL LIEPTAGVGV AAVLSQHFQT VSPEVKNICI VLSGGNVDLT SSITWVKQAE RPASYQSVSV |