Learn More
R&D Systems™ Recombinant Mouse Coagulation Factor Xa Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 6724-SE-010
Product Code. 18158600
  • £357.00 / 10µg
Explore more special offers
Description
Description
The Recombinant Mouse Coagulation Factor Xa Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Coagulation Factor Xa Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2 μm filtered solution in MES, NaCl and CaCl 2 .
29 kDa & 10 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
14058
Coagulation Factor X
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Coagulation Factor X protein Met1-Asn481,with a C-terminal 10-His tag. Accession &Num; O88947 Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa.
Recombinant Mouse Coagulation Factor X is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag.
Recombinant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Recombinant Mouse Coagulation Factor Xa Protein > 10ug; Carrier Free