missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MUC5AC Protein
A DNA sequence encoding the Mus musculus (Mouse) MUC5AC, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_034974.1 |
---|---|
Gene ID (Entrez) | 17833 |
Name | MUC5AC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077011
|
enQuireBio™
QP8891-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097011
|
enQuireBio™
QP8891-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087011
|
enQuireBio™
QP8891-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007021
|
enQuireBio™
QP8891-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017021
|
enQuireBio™
QP8891-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027021
|
enQuireBio™
QP8891-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_034974.1 | |
MUC5AC Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17833 | |
Research Use Only | |
Muc5ac | |
Recombinant Protein | |
E. coli | |
CPKNSSLIVT YEEGACCPTQ NCSSQKGCEV NGTLYQPGDV VSSSLCERCL CEVSSNPLSD VFMVSCETEL CNTQCPKGSE YQAMPGQCCG KCIPKTCPFK NNSGSTYFYQ PGELWAEPGN PCVTHKCEKF QDVLMVVTMK TECPKINCPQ GQAQLREDGC CYDCPLPNQQ KCTVHQRQQI IRQQNCSSEG PVSISYCQGN CGDSISMYSL EANKVEHTCE CCQELQTSQR NVTLRCDDGS SQTFSYTQVE KCGCLGQQCH ALGDTSHAES | |
Greater than 90% as determined by SDS-PAGE. |