missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Insulin Protein
A cDNA sequence encoding the Insulin was constructed and used to recombinantly synthesize the protein.
£122.00 - £1100.00
Specifications
Gene ID (Entrez) | 397415 |
---|---|
Name | Insulin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | INS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15915008
|
enQuireBio™
QP10733-1GR |
1 g |
£1100.00
1g |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925008
|
enQuireBio™
QP10733-250MG |
250 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15935008
|
enQuireBio™
QP10733-25MG |
25 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
397415 | |
Research Use Only | |
INS | |
Recombinant Protein | |
Porcine | |
MALWTRLLPL LALLALWAPA PAQAFVNQHL CGSHLVEALY LVCGERGFFY TPKARREAEN PQAGAVELGG GLGGLQALAL EGPPQKRGIV EQCCTSICSL YQLENYCN | |
Greater than 97.0% as determined by SEC-HPLC. |
Insulin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
>27 units/mg. | |
Porcine | |
Untagged | |
The Insulin Porcine was lyophilized from a concentrated (1 mg/ml) solution with no additives. |