missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein G - Sepharose™ 4B

Recombinant Protein G - Sepharose™ 4B

Brand:  Novex™ 101243

1824.70 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10405533

  • £2570.00 / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This product is prepared by covalently coupling highly purified rec-Protein G to Sepharose™ 4B. It is supplied as a 50% suspension containing rec-Protein G-Sepharose™ 4B + PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin affinity purification and Immunoprecipitation.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Protein G Sepharose
    Affinity
    This product is supplied as a 50% suspension (25 ml Recombinant Protein G-Sepharose™ 4B + 25 ml PBS) Wet Ice
    Suspension
    Sepharose™
    Protein G
    25 mL
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.