missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat ANPEP / APN / CD13 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) ANPEP / APN / CD13, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_112274.1 |
---|---|
Gene ID (Entrez) | 81641 |
Name | ANPEP / APN / CD13 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976709
|
enQuireBio™
QP5648-ec-10ug |
10 μg | |||||||
15916719
|
enQuireBio™
QP5648-ec-50ug |
50 μg | |||||||
15966709
|
enQuireBio™
QP5648-ec-100ug |
100 μg |
£560.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15996709
|
enQuireBio™
QP5648-ec-200ug |
200 μg | |||||||
15906719
|
enQuireBio™
QP5648-ec-500ug |
500 μg | |||||||
15986709
|
enQuireBio™
QP5648-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_112274.1 | |
ANPEP / APN / CD13 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
81641 | |
Research Use Only | |
Anpep | |
Recombinant Protein | |
E. coli | |
YAQEKNRNAE NSAIAPTLPG STSATTSTTN PAIDESKPWN QYRLPKTLIP DSYQVTLRPY LTPNEQGLYI FKGSSTVRFT CNETTNVIII HSKKLNYTNK GNHRVALRAL GDTPAPNIDT TELVERTEYL VVHLQGSLVK GHQYEMDSEF QGELADDLAG FYRSEYMEGG NKKVVATTQM QAADARKSFP CFDEPAMKAS FNITLIHPNN LTALSNMLPK DSRTLQEDPS WNVTEFHPTP KMSTYLLAYI VSEFKYVEAV SPNRVQIRIW ARPSAIDEGH GDYALQVTGP ILNFFAQHYN TAYPLEKSDQ IALPDFNAGA MENWGLVTYR ESALVFDPQS SSISNKERVV TVIAHELAHQ WFGNLVTVDW WNDLWLNEGF ASYVEFLGAD YAEPTWNLKD LIVLNDVYRV MAVDALASSH PLSSPANEVN TPAQISELFD SITY | |
Greater than 90% as determined by SDS-PAGE. |