missing translation for 'onlineSavingsMsg'
Learn More

Redox standard solution, Jenway™

Protection and care for pH electrodes

£198.00 - £460.00

Specifications

Quantity 500 mL
Chemical Name or Material pH Buffer
View More Specs

Products 2
Product Code Brand Solution Type Price Quantity & Availability  
Product Code Brand Solution Type Price Quantity & Availability  
11470609
View Documents
Jenway
90238-49
Redox Standard (200 mV)
£198.00
500mL
Please to purchase this item. Need a web account? Register with us today!
11480609
View Documents
Jenway
90238-50
Redox Standard (300 mV)
£460.00
500mL
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
Description

Description

  • Storage solution for pH electrodes
  • 500mL bottle
Specifications

Specifications

500 mL
pH Buffer
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Redox standard solution, Jenway™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.