Learn More
BINDER™ Reinforced Inner Chamber
Reinforced inner chamber for climate chambers and cooling incubation chambers.
Brand: BINDER™ 8012-2528
This item is not returnable.
View return policy
Description
- Reinforced inner chamber for bulky and heavy samples.
- Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
KBF PRO 1060 | |
Stainless Steel | |
Chamber Rack | |
200 kg |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BINDER™ Reinforced Inner Chamber > For use with KBF PRO 1060
Spot an opportunity for improvement?Share a Content Correction