missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ SP4 Glucose Broth
Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.
£51.88 - £691.00
Specifications
Description | SP4 Glucose Broth w/thallium acetate, penicillin |
---|---|
Product Type | Microbiology Media |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
13245509
|
Thermo Scientific™
R112585 |
SP4 Glucose Broth w/thallium acetate, penicillin; For isolation of Mycoplasma spp. |
£59.60
100mL |
In Stock
Log in to see stock. |
|||||
15436424
|
Thermo Scientific™
R20057 |
SP4 Glucose Broth w/thallium acetate, penicillin, 90mL |
£51.88
90mL |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
13225639
|
Thermo Scientific™
R20376 |
SP4 Glucose Broth; w/thallium acetate;D - 5mL; 20/Pk |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.
- Penicillin inhibits gram-positive bacteria
Specifications
SP4 Glucose Broth w/thallium acetate, penicillin | |
Microbiology Media |
REF R112585 not intended for IVD use.