Learn More
Electrothermal™ Replacement Digi-Mantle Heater Cartridge
Enables a round bottom flask to be stirred and heated up to an element temperature of 450°C. Electrothermal™ Replacement Digi-Mantle Heater Cartridge uses precise electronic control to provide accurate e of temperature and stirring measures.
Brand:  Electrothermal™ 36204-09
 View more versions of this product
Product Code. 11777873
  • £870.00 / Each
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with the OMCA Digital Controller.
  • Stir and heat round bottom flasks up to an element temperature of 450°C
  • Uses precise electronic control to give an accurate measure of temperature and stirring.
  • Modular design allows heating cartridges of different sizes to be interchanged using the same OMCA digital controller as a base
  • Cost-effective and versatile
Specifications
Specifications
1000 mL
OMCA Digi-Mantle
Replacement Heating Cartridge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Electrothermal™ Replacement Digi-Mantle Heater Cartridge >