missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Resazurin Tablets, Technical, Fisher Chemical™
£31.60 - £165.00
Chemical Identifiers
CAS | 550-82-3 |
---|---|
Molecular Formula | C12H7NO4 |
Molecular Weight (g/mol) | 229.191 |
MDL Number | 66739 |
InChI Key | PLXBWHJQWKZRKG-UHFFFAOYSA-N |
PubChem CID | 11077 |
ChEBI | CHEBI:8806 |
IUPAC Name | 7-hydroxy-10-oxidophenoxazin-10-ium-3-one |
SMILES | C1=CC2=C(C=C1O)OC3=CC(=O)C=CC3=[N+]2[O-] |
Description
Chemical Identifiers
550-82-3 | |
229.191 | |
PLXBWHJQWKZRKG-UHFFFAOYSA-N | |
CHEBI:8806 | |
C1=CC2=C(C=C1O)OC3=CC(=O)C=CC3=[N+]2[O-] |
C12H7NO4 | |
66739 | |
11077 | |
7-hydroxy-10-oxidophenoxazin-10-ium-3-one |
Specifications
550-82-3 | |
66739 | |
C1=CC2=C(C=C1O)OC3=CC(=O)C=CC3=[N+]2[O-] | |
229.191 | |
CHEBI:8806 | |
Orange to Red |
C12H7NO4 | |
PLXBWHJQWKZRKG-UHFFFAOYSA-N | |
7-hydroxy-10-oxidophenoxazin-10-ium-3-one | |
11077 | |
Glass Jar | |
Solid |