Learn More
Restek™ HPLC 30-Column Storage Cabinet
Saves space and protects columns.
Brand: Restek™ 25159

Specifications
HPLC column storage Cabinet | |
Hinged door is clear for quick identification of column labels or tags | |
HPLC columns (min. 100mm L) | |
Hinged door | |
PVC |
17 x 15 x 3 in. | |
Easy-to-install cabinet saves space and protects columns, the hinged door is clear to allow quick identification of column labels or tags | |
30 columns (minimum 100 mm L) | |
Easy-to-install, wall-mountable (screws not included) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.