Learn More
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R
Eppendorf Rotor for Centrifuge 5910 Ri and 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds, buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
Brand:  Eppendorf™ 5895201008
1477.25 GBP valid until 2025-12-31
Use promo code "24378" to get your promotional price.
 View more versions of this product
Product Code. 15873212
  • £1535.00 / Each
Explore more special offers
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor
4,347 x g
Centrifuges 5910 Ri and 5920 R
4
4 x 1,000 mL
4,500 rpm
Without Buckets
S-4xUniversal
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R > Description: S-4xUniversal - without buckets, Capacity: 4 x 1,000 mL, Max. RCF: 4,347 x g, Max. Speed: 4,500 rpm