Learn More
Fisherbrand™ Solid Rubber Stoppers
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
Brand: Fisherbrand™ RB003/S
Description
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
- Temperature range -40° to +70°C
- Not suitable for use with petroleum oils and hydrocarbon solvents
Specifications
Laboratory use | |
50/Pk. | |
3 mm | |
Rubber | |
3 |
12mm | |
Red | |
4.5 mm | |
Solid Stopper |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.