Learn More
Rubidium chloride, 99% (metals basis), Thermo Scientific Chemicals
Description
It is used in testing for perchloric acid in laboratories, and is a source of rubidium. Rubidium chloride is used as a catalyst and additive in petrol. It has been employed to prepare molecular nanowires for nanoscale devices. Thin layer of rubidium chloride enhance the attributes of organic light-emitting diodes. Rubidium chloride also plays an essential role for enhancement in the memory storage property. It is an excellent non-invasive biomarker, and used in biomedical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1390°C |
Melting Point | 718°C |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 99% (metals basis) |
Merck Index | 14,8284 |
Solubility Information | Soluble in water; Practically insoluble in alcohol. |
IUPAC Name | rubidium(1+);chloride |
Formula Weight | 120.92 |
Percent Purity | 99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U14K024 | Certificate of Analysis | 07/11/2024 | 012892.14,012892.06,012892.22 |
R28K002 | Certificate of Analysis | 29/07/2024 | 012892.14,012892.06,012892.22 |
P21K001 | Certificate of Analysis | 04/04/2024 | 012892.14,012892.06,012892.22 |
P07I040 | Certificate of Analysis | 27/12/2023 | 012892.14,012892.06,012892.22 |
R23I052 | Certificate of Analysis | 17/11/2022 | 012892.14,012892.06,012892.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.