Learn More
Rubidium chloride, 99% (metals basis), Thermo Scientific Chemicals
Testing for perchloric acid, source of rubidium, catalyst
£40.24 - £544.71
Chemical Identifiers
CAS | 7791-11-9 |
---|---|
Molecular Formula | ClRb |
Molecular Weight (g/mol) | 120.92 |
MDL Number | MFCD00011187 |
InChI Key | FGDZQCVHDSGLHJ-UHFFFAOYSA-M |
Synonym | rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chlorideShow More |
PubChem CID | 62683 |
ChEBI | CHEBI:78672 |
SMILES | [Cl-].[Rb+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15421027
|
Thermo Scientific Alfa Aesar
012892.06 |
5 g |
£40.24
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377627
|
Thermo Scientific Alfa Aesar
012892.14 |
25 g |
£175.88
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367627
|
Thermo Scientific Alfa Aesar
012892.22 |
100 g |
£544.71
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in testing for perchloric acid in laboratories, and is a source of rubidium. Rubidium chloride is used as a catalyst and additive in petrol. It has been employed to prepare molecular nanowires for nanoscale devices. Thin layer of rubidium chloride enhance the attributes of organic light-emitting diodes. Rubidium chloride also plays an essential role for enhancement in the memory storage property. It is an excellent non-invasive biomarker, and used in biomedical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7791-11-9 | |
120.92 | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
62683 | |
[Cl-].[Rb+] |
ClRb | |
MFCD00011187 | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chlorideShow More | |
CHEBI:78672 |
Specifications
1390°C | |
7791-11-9 | |
Crystalline | |
ClRb | |
14,8284 | |
Soluble in water; Practically insoluble in alcohol. | |
[Cl-].[Rb+] | |
120.92 | |
CHEBI:78672 | |
99% | |
1.493 | |
2.800 g/mL | |
Rubidium chloride |
718°C | |
White | |
99% (metals basis) | |
MFCD00011187 | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
rubidium(1+);chloride | |
62683 | |
120.92 | |
Odorless | |
Hygroscopic | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 232-240-9
missing translation for 'rtecsNumber' : VL8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.