missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Benzyl-2-oxazolidinone, 99%, Thermo Scientific Chemicals

£19.40 - £218.09

Chemical Identifiers

CAS 90719-32-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00064496
InChI Key OJOFMLDBXPDXLQ-VIFPVBQESA-N
Synonym s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
PubChem CID 736225
IUPAC Name (4S)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@@H](CC2=CC=CC=C2)CO1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11433767
View Documents
Thermo Scientific Alfa Aesar
A18236.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11443767
View Documents
Thermo Scientific Alfa Aesar
A18236.06
5 g
£63.11
5g
Please to purchase this item. Need a web account? Register with us today!
11453767
View Documents
Thermo Scientific Alfa Aesar
A18236.14
25 g
£218.09
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(S)-(-)-4-Benzyl-2-oxazolidinone is used in chiral auxiliary asymmetric alkylation. It has been used in the synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides. It was used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-4-Benzyl-2-oxazolidinone is used in chiral auxiliary asymmetric alkylation. It has been used in the synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides. It was used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

90719-32-7
177.20
OJOFMLDBXPDXLQ-VIFPVBQESA-N
736225
O=C1N[C@@H](CC2=CC=CC=C2)CO1
C10H11NO2
MFCD00064496
s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
(4S)-4-benzyl-1,3-oxazolidin-2-one

Specifications

87°C to 89°C
C10H11NO2
3649667
Insoluble in water.
O=C1N[C@@H](CC2=CC=CC=C2)CO1
177.20
177.2
Hygroscopic
(S)-(-)-4-Benzyl-2-oxazolidinone
90719-32-7
MFCD00064496
s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
OJOFMLDBXPDXLQ-VIFPVBQESA-N
(4S)-4-benzyl-1,3-oxazolidin-2-one
736225
−64° (c=1 in Chloroform)
99%
Videos
Safety and Handling

Safety and Handling

P261-P262-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P314-P330-P332+P313-P362-P501c

H302-H315-H320-H335-H373

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.