Learn More
Brady™ SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
The ultimate locks for all of your safety and security needs
£19.15 - £114.00
Specifications
Product Type | Keyed Padlocks |
---|---|
Key Type | Keyed Different |
Material | Nylon |
Diameter (English) Shackle | 0.25 in. |
Diameter (Metric) Shackle | 6.4 mm |
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | |||||
17170281
|
Brady™
150230 |
1 Ea. | Orange |
£19.15
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17180281
|
Brady™
150231 |
1 Ea. | Black |
£19.15
Each |
In Stock
Log in to see stock. |
|||||
17190281
|
Brady™
150232 |
1 Ea. | Yellow |
£19.15
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17180311
|
Brady™
150272 |
1 Ea. | Purple |
£19.15
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17190311
|
Brady™
150273 |
1 Ea. | Green |
£19.15
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17170351
|
Brady™
150318 |
1 Ea. | Brown |
£19.15
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17140371
|
Brady™
150342 |
1 Ea. | Red |
£19.15
Each |
In Stock
Log in to see stock. |
|||||
17190381
|
Brady™
150365 |
1 Ea. | White |
£19.55
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17100391
|
Brady™
150366 |
1 Ea. | Blue |
£19.15
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17100281
|
Brady™
|
6 Pk. | Blue |
£114.00
Pack of 6 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17120311
|
Brady™
150265 |
6 Pk. | Yellow |
£114.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
17190341
|
Brady™
150308 |
6 Pk. | White |
£114.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
17100351
|
Brady™
150309 |
6 Pk. | Brown |
£114.00
Pack of 6 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
17110351
|
Brady™
150310 |
6 Pk. | Orange |
£114.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
17120351
|
Brady™
150311 |
6 Pk. | Red |
£114.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
17190361
|
Brady™
150334 |
6 Pk. | Green |
£114.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
17110381
|
Brady™
150350 |
6 Pk. | Purple |
£114.00
Pack of 6 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17120381
|
Brady™
|
6 Pk. | Black |
£114.00
Pack of 6 |
In Stock
Log in to see stock. |
|||||
Description
- Compliments the Brady™ SafeKey Padlock range and have the same features
- The new designed and innovative keying mechanism provides you much more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout
- The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted
- The lock mechanism has 6 precision steel tumbler blades for a high number of key differences and tamper resistance
- Locks are lightweight and resist impact, temperature, chemicals and corrosion
- Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling
- The colour-matched key and lock body allows a fast identification in the application
- 1 key supplied with each lock
- Key retaining (Key won’t release until the padlock shackle is securely closed)
- Custom keying, engraving, charting, Master Key and Grand Master Key are available
Specifications
Keyed Padlocks | |
Nylon | |
6.4 mm | |
38 mm | |
6 | |
1.3 in. | |
33 mm |
Keyed Different | |
0.25 in. | |
1.5 in. | |
Plastic | |
1.5 in. | |
38 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brady™ SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different