missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT Safety Spouts
Safety pourer with integrated screw cap for safe dispensing of liquids.
£16.70 - £223.00
Specifications
For Use With (Equipment) | Canister, Container & Bottles |
---|---|
Product Type | Safety Spout |
Product Code | Brand | Closure Size | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Description | Price | Quantity & Availability | |||||
10399309
|
SCAT
610500 |
S55 | Rigid |
£17.15
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10589680
|
SCAT
610499 |
S55 | Flexible |
£16.70
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10309509
|
SCAT
610504 |
S60/61 | Rigid with Vent |
£223.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10262770
|
SCAT
610501 |
S60/61 | Flexible |
£17.60
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
|||||
10639880
|
SCAT
610502 |
S60/61 | Fixed |
£16.70
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10344470
|
SCAT
610503 |
S70/71 | Flexible |
£16.70
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
Description
Specifications
Canister, Container & Bottles | |
Safety Spout |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCAT Safety Spouts