missing translation for 'onlineSavingsMsg'
Learn More
Salicylamide, 99%, Thermo Scientific Chemicals Product Code.: 10236430

Salicylamide, 99%, Thermo Scientific Chemicals

Product Code. 10236430
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10236430

Brand: Thermo Scientific Acros 229360050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-45-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007978
InChI Key SKZKKFZAGNVIMN-UHFFFAOYSA-N
Synonym salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevinShow MoreShow Less
PubChem CID 5147
ChEBI CHEBI:32114
IUPAC Name 2-hydroxybenzamide
SMILES NC(=O)C1=CC=CC=C1O

Specifications

Melting Point 139.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on anhydrous substance)
Packaging Glass Bottle
Linear Formula C6H4(OH)CONH2
Quantity 5 g
Beilstein 10, 87
Merck Index 15, 8465
Solubility Information Solubility in water: soluble in hot water. Other solubilities: 0.2% in water (30°C), 10.0% in propylene glycol (5°C), soluble in alcohol, chloroform and ether, slightly soluble in carbon tetrachloride
Formula Weight 137.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Salicylamide, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0354188 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number229360050,229360010,229362500
Lot NumberA0446088 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog Number229360050,229360010,229362500
Lot NumberA0384848 Certificate TypeCertificate of Analysis Date20/05/2022 Catalog Number229360050,229360010,229362500
Lot NumberA0436640 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number229360050,229360010,229362500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Salicylamide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.