Learn More
Thermo Scientific™ Sample Cuvettes for Eutech™ TB 1000 Turbidity Meter
The CyberScan TB1000 series turbidity meters and cuvettes are designed to meet the stringent requirements of today's scientists.
Brand:  Thermo Scientific™ 01X265701
Product Code. 12911778
  • £116.12 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Sample Cuvettes for Eutech™ TB 1000 Turbidity Meter are borosilicate glass vials with light-shield screwcaps, fill line and indexing marks.The cuvettes come in packs of three.
Features:
  • Pack of 3 sample cuvettes for TB1000W (white light turibidity meter) & TB1000IR( infra-red turbidity meter)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CyberScan TB 1000
Sample Cuvettes
3 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Sample Cuvettes for Eutech™ TB 1000 Turbidity Meter >