Learn More
Velp Scientifica™ Sample Racks for Digestion Units
Facilitates nitrogen analysis and protein determination. Velp Scientifica™ Sample Racks for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
Brand: Velp Scientifica™ A00000172
This item is not returnable.
View return policy
Description
- Available with or without heat shields
- Available to fit a number of tube sizes and counts
Specifications
Sample Rack | |
DKL12 |
Product Suggestions
Customers who viewed this item also viewed.
VEL055 VELP DKL 21
File size: 817.4 kB
Water analysis
File size: 7.3 MB
Product Content Correction
Product Title
Velp Scientifica™ Sample Racks for Digestion Units > For Use With: DKL12, 250 ml tubes
Spot an opportunity for improvement?Share a Content Correction