Learn More
Velp Scientifica™ Sample Racks for Digestion Units
Facilitates nitrogen analysis and protein determination. Velp Scientifica™ Sample Racks for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
Brand: Velp Scientifica™ A00000246
This item is not returnable.
View return policy
Description
- Available with or without heat shields
- Available to fit a number of tube sizes and counts
Specifications
Sample Rack | |
DKL 20 |
Product Content Correction
Product Title
Velp Scientifica™ Sample Racks for Digestion Units > For Use With: 400 ml test tubes
Spot an opportunity for improvement?Share a Content Correction