Learn More
Sampling Systems™ Powder Lance
Versatile sampler that can be used horizontally through a sack or vertically to take samples from drums, kegs, super sacks, and more. Sampling Systems™ Powder Lance is a cross sectional bulk sampler for cohesive powders that can be used to take large volume samples of cohesive powders.
Brand: Sampling Systems™ 2250A-2000
Description
- Manufactured in 316L Stainless Steel
- Multilevel discreet samples
Specifications
Powder Lance | |
360 mL | |
1600 mm |
Stainless Steel (316L) | |
2030 mm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.