Learn More
Thermo Scientific™ Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
Brand:  Thermo Scientific™ BRE0006591
 View more versions of this product
Product Code. 16373348
  • £181.00 / Each
Explore more special offers
Description
Description
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
  • Sample cones made from nickel and platinum
  • Skimmer cones made from nickel and platinum
  • Platinum, boron free skimmer cone (for semiconductor-related applications)
  • Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifications
Specifications
Skimmer Cone Insert 4,5 (Robust Interface)
Skimmer Cone Insert 4.5 (Robust Plasma)
iCAP Q/Qnova Series ICP-MS Systems
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems >