missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals SANT-1 Product Code.: 15436819

Thermo Scientific Chemicals SANT-1

Product Code. 15436819
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy

Product Code. 15436819

Brand: Thermo Scientific Chemicals J65294.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling; high affinity antagonist of smoothened activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling
  • High affinity antagonist of smoothened activity
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description Pale Yellow
Content And Storage -30°C to -10°C
For Use With (Application) Potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling; high affinity antagonist of smoothened activity
CAS 304909-07-7
Molecular Formula C23H27N5, C23H27N5
Formula Weight 373.49g/mol
Synonym (4-Benzyl-piperazin-1-yl)-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-ylmethylene)-amine
Sensitivity Moisture and Light Sensitive
Solubility Soluble in DMSO with warming
Product Type Signal Transduction Reagent
Form Solid
InChI Key FOORCIAZMIWALX-UHFFFAOYSA-N
SMILES CC1=NN(C(C)=C1C=NN1CCN(CC2=CC=CC=C2)CC1)C1=CC=CC=C1
IUPAC Name N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine
Molecular Weight (g/mol) 373.50
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU19K051 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberJ65294.MA
Lot NumberT11K031 Certificate TypeCertificate of Analysis Date29/07/2024 Catalog NumberJ65294.MA
Lot NumberP01K021 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog NumberJ65294.MA
Lot NumberN14J060 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberJ65294.MA
Lot NumberU22J024 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberJ65294.MA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals SANT-1 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.