Learn More
Sartorius Sartoclear Dynamics™ Lab V Filtration Kit
Sartorius Sartoclear Dynamics™ Lab V Kits designed for the clarification and sterile filtration of mammalian cell culture with diatomaceous earth as filter aid.
Brand:  Sartorius SDLV-0500-10E0-2
Additional Details : CAS Number : 91053-39-3 Weight : 3.50000kg
 View more versions of this product
Product Code. 17444432
  • £795.00 / Pack of 24
Estimated Shipment: 03-09-2024
to see stock.
Add to basket
Description
Description
Comprised of:
  • A graduated funnel with a polyethersulfone (PES) membrane, a vacuum adapter and a lid
  • A bottle, with graduation and writing field
  • A tube connector for vacuum connection (for stand-alone filtration)
  • A screw cap for storage of the filtrate (individually wrapped to maintain sterility) history
Benefits:
  • Kits for a single-step mammalian cell culture harvesting
  • Ready to use
  • Fast and effortless
Specifications
Specifications
Vacuum Filtration Units
2 x 180E04---------E (24 x Sartolab™ RF 500; 0.22 μm; PES, single-packaged; sterile); 1 x SDLKG-10.0------2 (24 x pouches of filter aid, 10 g)
500 mL