Learn More
Sartorius Vivapure™ Q Mini H IEX Spin Columns

Ready-to-use centrifugal devices based on Sartobind Membrane Adsorber technology as their chromatography matrix
Brand: Sartorius VS-IX01QH24
242.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Vivapure Mini Ion Exchange (IEX) spin columns are centrifugal devices, incorporating Sartobind Membrane Adsorber technology as their chromatography matrix. Vivapure IEX spin columns make protein purification as easy as filtration. The devices are ready-to-use and do not bear the risk of running dry. For many protein purification applications, they can replace time-consuming and tedious column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel. VIvapure Mini is available in Quaternary ammonium (Q) membrane chemistry.
Typical Applications
- Sample preparation prior to 1-D or 2-D PAGE
- Purification scouting of an unknown protein
- Removal of endotoxins from monoclonal antibodies
- Preparation of heme moiety from heme containing protein for functional analysis
- Polishing of His-tagged proteins
- General protein purification
- Detergent removal from protein solutions
- Purification of antibodies from serum, ascites or tissue culture supernatant
- Establishing a purification/ pre-purification protocol for a given protein
- HPLC/FPLC sample preparation
- Purification of membrane-bound proteins
- Fast centrifugal spin columns
- Easy and ready-to-use
- Binding capacity 1mg up to 4 mg protein
- Ideal for parallel processing

Specifications
240 μL | |
Polypropylene | |
Stabilized regenerated Cellulose | |
Sample fractionation, purification condition scouting, small scale purification | |
24 x 0.5 mL | |
Protein Purification |
0.4 mL | |
3 to 5 μm | |
24/Pk. | |
Spin Column | |
Quaternary Ammonium | |
Ion Exchange Spin Column |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.