Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
Brand: Thermo Scientific™ RH10-15
This item is not returnable.
View return policy
Description
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Centrifuge Tubes: 10 x 15mL conical, Medium capacity models | |
SPD1030, SPD120, SPD130, SPD140DDA | |
RH10-15 |
Rotor | |
Centrifuge Tubes | |
Traditional |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators > Centrifuge Tubes: 10 x 15mL conical, Medium capacity models
Spot an opportunity for improvement?Share a Content Correction