missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals SB 203580, 98% Product Code.: 15484079

Thermo Scientific Chemicals SB 203580, 98%

Product Code. 15484079
50 mg
Click to view available options
Quantity:
25 mg
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 15484079

Brand: Thermo Scientific Chemicals J61482.MF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152121-47-6
Molecular Formula C21H16FN3OS, C21H16FN3OS
Synonym 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole

Specifications

Quantity 50 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Highly specific new inhibitor of p38 kinase and MAP Kinase homologues
Formula Weight 377.44g/mol
Melting Point 235°C to 247°C
Product Type Signal Transduction Reagent
Form Powder
InChI Key CDMGBJANTYXAIV-UHFFFAOYNA-N
SMILES CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
IUPAC Name 4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine
Molecular Weight (g/mol) 377.44
Assay 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY25K027 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberJ61482.MC,J61482.MB,J61482.MF
Lot NumberY11K005 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ61482.MC,J61482.MB,J61482.MF
Lot NumberW05K033 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberJ61482.MC,J61482.MB,J61482.MF
Lot NumberT24K025 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog NumberJ61482.MC,J61482.MB,J61482.MF
Lot NumberM11K045 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog NumberJ61482.MC,J61482.MB,J61482.MF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals SB 203580, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.