missing translation for 'onlineSavingsMsg'
Learn More
Scandium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11366268

Scandium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11366268
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11366268

Brand: Thermo Scientific Alfa Aesar 011221.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107552-14-7
Molecular Formula N3O9Sc
Molecular Weight (g/mol) 230.968
MDL Number MFCD00011220
InChI Key DFCYEXJMCFQPPA-UHFFFAOYSA-N
Synonym scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium saltShow MoreShow Less
PubChem CID 166818
IUPAC Name scandium(3+);trinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sc+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 1 g
UN Number UN1477
Merck Index 14,8392
Solubility Information Soluble in water and alcohol.
Formula Weight 230.97 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM13L041 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number011221.03,011221.06
Lot NumberW16K004 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number011221.03,011221.06
Lot NumberT15K003 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number011221.03,011221.06
Lot NumberN05K048 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number011221.03,011221.06
Lot NumberU08J040 Certificate TypeCertificate of Analysis Date13/10/2023 Catalog Number011221.03,011221.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.