Learn More
3M™ Scotch™ Fine Line Tape
3M™ Scotch™ Fine Line Tape is a high-performance polypropylene plastic tape that has both a low profile and high adhesion for use in painting long straight lines and sweeping curves where a crisp, sharp paint line is desired.
Brand: 3M™ 7000048455
This item is not returnable.
View return policy
Description
- Thin tape provides excellent color line separation
- Proven excellent for painting long straight lines and smooth curves
- Resistant to most solvents and moisture
- Often used as a layout tape for high value multi-color and custom painted graphics.
Specifications
Green | |
Fine Line Tape | |
218 | |
5 mil |
Rubber | |
Masking for Painting, Paint Preparation Masking | |
144 Rolls/Cs. (12 Rolls/Pk. 12 Pk./Cs.) | |
60 yd. x 1/8 in. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Product Title
3M™ Scotch™ Fine Line Tape > Green
Spot an opportunity for improvement?Share a Content Correction