Learn More
3M™ Petrifilm™ Select E. Coliplate
Specific for E. coli, only E. coli colonies are visible which numerate within 24 hours
Brand:  3M™ Petrifilm™ 7100126814
Encompass_Preferred
 View more versions of this product
Product Code. 17233784
  • £79.11 / Pack of 50
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specific test for E. coli, only E. coli colonies are visible
  • E. coli enumerates in just 24 hours
  • Contains a ß-glucuronidase indicator
Specifications
Specifications
E. coli
25 Pack