Learn More
Brady™ Self-laminating Cryogenic Polyester Labels
Securely label lab tubes and vials for cryogenic storage with this self-laminating, ultra-thin polyester label material (B-461). Print almost anywhere with a compact Brady™ portable label printer.
Brand:  Brady™ 170878
 View more versions of this product
Product Code. 17831719
  • £65.20 / Pack of 180
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws and slides
  • Able to withstand extreme temperatures from -196°C to 130°C (-320°F to 266°F), immersion in liquid nitrogen and autoclave use
  • Resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% Clorox solution
  • Made with a clear overlaminate that protects the print and maintains content visibility
Specifications
Specifications
Autoclavable
M5-125-461
Black on White/Transparent
Polyester
Laboratory Labeling
180 Labels/Cartridge
0.0019 in.
1 in.
4.5 cm
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Die-cut
Labels
BMP51, M511
-321°F to +230°F
1 x 1.75 in.
1.75 in.
0.5 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Self-laminating Cryogenic Polyester Labels > Self-laminating Cryogenic Polyester Labels; Color: Black on White/Transparent; Dimensions (LxW): 1 x 1.75 in.; For Use With (Equipment): BMP51, M511; Format: Die-cut