Learn More
Ansell™ Sensilite™ 48-135 Series White and Gray Polyurethane Light Weight Gloves Product Code.: 15240009
Add to basket
Ansell™ Sensilite™ 48-135 Series White and Gray Polyurethane Light Weight Gloves
Product Code. 15240009 Shop All Ansell Products
8
Click to view available options
Size:
6
7
8
9
10
11
Product Code. 15240009
Brand: Ansell™ 48135080
Add to basket
Add to basket
Meets most electrostatic discharge protection needs and provides flexibility and dexterity.
  • Special purpose gloves
  • 15-gauge nylon liner
  • Added carbon fibre offers economical product-protection solution
  • Comfort and good fit are assured through seamless construction and a wide range of sizes
  • With the palm dipped in a polyurethane coating, offers good mechanical resistance and durability
  • Fingertip-dipped version allows better fit and flexibility
  • Coated fingertip also prevents the transfer of perspiration to the electronic part
Applications:
  • Final assembly of dashboard, cables and wires
  • Final assembly of electronic components and mother boards
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 113X
Cuff Material Polyurethane
Coating Material Polyurethane
Product Type Lightweight Gloves
Size 8
Style 48-135
Finger Style Coated-fingertip
Glove Cuff Style Knit
Lining Nylon Carbon Fiber
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Good mechanical resistance
Series 48-135
Product Title
Ansell™ Sensilite™ 48-135 Series White and Gray Polyurethane Light Weight Gloves >