missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G-25 Media

Use of Sephadex™ is well established as a gel filtration medium for desalting and buffer exchange in industrial applications. Cytiva Life Sciences™ Sephadex™ G-25 Media quickly desalts, removes contaminants and transfers to a new buffer in a single step.

Brand:  Cytiva 17-0033-01

 View more versions of this product

Product Code. 11580774

  • £306.50 / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Established medium for desalting and buffer exchange

  • Gel filtration medium
  • Available in coarse, medium, fine and superfine grades
  • All grades satisfy process-scale chromatography requirements
  • Available in lab packs or prepacked columns

Specifications:

  • Working pH range: 2 to 13
  • Bed volume (in distilled water): 4 to 6mL/g
  • Fractionation range dextrans/globular proteins: 1× 102 to 5 × 103 / 1 × 103 to 5 × 103 Mr
  • Max. flowrate/operating pressure: the beads behave as rigid spheres obeying Darcy's Law

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 g
38 to 235 μm (Wet Form), >50 μm (Dry Form)
Sephadex G-25 Media
2.6cm
11.81 in.
>100 cm/h, pressure drop cm H2O/bed height=2, bed height 30 cm, 2.6 cm i.d.
Cross-linked Dextran
>100 cm/hr.
4 to 6mL/g in Distilled Water
Solid Spherical Bead
Desalting and Buffer exchange and removal free label in industrial applications
30cm
2 to 13
4 to 30°C, dry resins. Used resins 4 to 8 °C in 20% Ethanol or 0.01 M NaOH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Sephadex™ G-25 Media > Medium; Particle size: 38 to 235μm; 100g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.