missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sepharose Agarose Gel Filtration Media
Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.
Brand: Cytiva 17-0110-01
This item is not returnable.
View return policy
Description
European Pharmacopoeia Solutions
- Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
- All are manufactured and tested in compliance with the Ph. Eur.
- Supplied with a Certificates of Analysis
- Lot No. and expiry date are stated on all product labels
Order Info
Supplied in 1L plastic containers
Specifications
1L | |
45 to 165 μm | |
Solid Bead | |
Allow characterization or cleaning up of samples containing components of diverse molecular weight | |
10cm | |
4 to 9 | |
4 to 30°C, 20% Ethanol |
<14 cm/hr. | |
Sepharose Agarose Gel Filtration Media | |
5cm | |
3.94 in. | |
100-200 cm/h, pressure drop cm H2O/bed height=15, bed height 10 cm, 5 cm i.d. | |
6% Agarose |