missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sepharose Agarose Gel Filtration Media
Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.
£675.49 - £872.00
Specifications
Quantity | 1 L |
---|---|
Product Type | Sepharose Agarose Gel Filtration Media |
Format | Solid Bead |
For Use With (Application) | Allow characterization or cleaning up of samples containing components of diverse molecular weight |
Product Code | Brand | Flow Rate | Particle Size | pH Range | Support Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Particle Size | pH Range | Support Type | Price | Quantity & Availability | ||
10004634
|
Cytiva
17-0150-01 |
<26 cm/hr. | 45 to 165 μm | 3 to 13 | 2% Agarose |
£872.00
1 L |
In Stock
Log in to see stock. |
||
10217754
|
Cytiva
17-0140-01 |
<15 cm/hr. | 60 to 200 μm | 3 to 13 | 2% Agarose |
£836.00
1 L |
In Stock
Log in to see stock. |
||
10605355
|
Cytiva
17-0120-01 |
<11.5 cm/hr. | 45 to 165 μm | 4 to 9 | 4% Agarose |
£815.00
1 L |
In Stock
Log in to see stock. |
||
10359864
|
Cytiva
17-0130-01 |
<10 cm/hr. | 60 to 200 μm | 4 to 9 | 2% Agarose | N/A | |||
10702627
|
Cytiva
17-0160-01 |
<30 cm/hr. | 45 to 165 μm | 3 to 12 | 6% Agarose |
£872.00
1 L |
In Stock
Log in to see stock. |
||
10695345
|
Cytiva
17-0110-01 |
<14 cm/hr. | 45 to 165 μm | 4 to 9 | 6% Agarose |
£815.00
1 L |
In Stock
Log in to see stock. |
||
Description
- Bead-formed agarose-based gel filtration media
- Proven base matrix for coupling affinity ligands
- Good separation over a wide molecular weight range
Sepharose gels
- Available in three different formulations: 2% (2B), 4% (4B),and 6% (6B) agarose
- Working pH range: 4-9
- Cross-linked derivatives of Sepharose gels, with same agarose concentrations
- Working pH range: 3-13
- Greater chemical and physical resistance
- Equivalent selectivity, better flow characteristics
- Ideal for separations in organic solvents
Order Info
Supplied in 1L plastic containers
Specifications
1 L | |
Sepharose Agarose Gel Filtration Media | |
Solid Bead | |
Allow characterization or cleaning up of samples containing components of diverse molecular weight |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title