missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sepharose Agarose Gel Filtration Media

Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.

Brand:  Cytiva 17-0160-01

793.86 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Product Code. 10702627

  • £872.00 / 1 L
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

European Pharmacopoeia Solutions
  • Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
  • All are manufactured and tested in compliance with the Ph. Eur.
  • Supplied with a Certificates of Analysis
  • Lot No. and expiry date are stated on all product labels

Order Info

Supplied in 1L plastic containers

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 L
45 to 165 μm
Solid Bead
Allow characterization or cleaning up of samples containing components of diverse molecular weight
10cm
6% Agarose
<30 cm/hr.
Sepharose Agarose Gel Filtration Media
5cm
3.94 in.
3 to 12
4 to 30°C, 20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • sepharose CL-6B

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers