Learn More
Cytiva Sera-Mag™ Streptavidin Magnetic Bead, medium biotin binding capacity
Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.
Brand: Cytiva 30152104011150
125.22 GBP valid until 2024-12-31
Use promo code "21948" to get your promotional price.
Code : LE
Additional Details : Weight : 0.13610kg
Description
Biotin Binding Capacity of 3500 to 4500pmol/mg beadsSpecifications
1% solids (10mg/mL) | |
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
1 μm |
2°C to 8°C | |
1 mL |