Learn More
Biochrom Serial Cable 9 Way M-F
Serial Cable 9 Way M-F for Use with Novaspec Plus/III, Libra S4/6, WPA S800/1200 Visible Spectrophotometers.
Brand:  Biochrom 80-2118-18
Product Code. 11345020
  • £43.00 / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ready to use
Specifications
Specifications
Serial Cable 9 Way M-F
1 Ea.
0/5 V
Biochrom Novaspec Plus/III, Libra S4/6, WPA S800/1200 Visible Spectrphotometers
Cable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Biochrom Serial Cable 9 Way M-F > Serial Cable 9 Way M-F