missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor
GREENER_CHOICEBIOPHARMA

The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. It is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.

Brand:  BINDER™ 9640-0009

8288.48 GBP valid until 2025-12-31
Use promo code "25039" to get your promotional price.


 View more versions of this product

Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 17303703

  • £10820.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Optimized double-pan humidification system with condensation protection
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data recording, can be read out in open format via USB interface
  • Fault diagnosis system with visual and acoustic alarm
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures
Specifications
Show More
Product Suggestions

Product Suggestions

Videos

Videos

  • Binder CO2 incubator CB170
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Binder CO2 incubator CB170

    SDS
    Documents Certificates
    View accessory products
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor > Model: CB170-230V, Capacity: 170 L, Voltage: 200/230 V, Frequency: 50/60 Hz

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.