Learn More
Mettler Toledo™ SevenGo Duo™ Pro pH/Ion/Conductivity Multi Parameter Meters
Portable instruments for pH, conducitivity, ion and dissolved oxygen determination
Brand:  Mettler Toledo™ 51302625
2095.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10627245
  • £2225.00 / Each
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The instruments are suited for mobile measurements both in the field and in the production environment. The SG23 SevenGo Duo™ pH/mV/conductivity meter is best suited for parallel routine measurements in dual-channel mode. In addition, it also offers Intelligent Sensor Management (ISM™). The SG78 and SG68 SevenGo Duo Pro™ instruments allow simultaneous determination and display of two electrochemical parameters with highest possible accuracy. They have the same comprehensive GLP functions as their SevenGo Pro™ counterparts. In addition, the SevenGo Duo pro™ instruments have extra security functions, such as Intelligent Sensor Management (ISM ™), PIN-protected device logon and PIN-protected system setting.
  • Portable instruments for pH, conductivity, ion and dissolved oxygen determination
  • Robust, watertight single- and dual-channel instruments for use in demanding conditions
  • Unique ergonomics and extremely simple operation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.01 μS/cm to 1000 mS/cm
0.01 to 100.0 Mohm/cm
0.1 mg/L to 600 g/L
Electrode Kit with InLab Expert Pro-ISM, InLab738-ISM (both IP67, 1.8m cable)
Electrode Kit
-2 to 20
0 to 80 psu
-5°C to +130°C (pH ATC), -30°C to +130°C (MTC)
SG78-USP
-1999 to 1999 mV
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ SevenGo Duo™ Pro pH/Ion/Conductivity Multi Parameter Meters > Model SG78-USP; Includes Electrode Kit with InLab Expert Pro-ISM, InLab738-ISM (both IP67, 1.8m cable)