Learn More
Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, Holds No. 20-25 Blades
Securely hold blades with the Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, constructed of #4 stainless steel.
Brand:  Epredia™ 5334
Product Code. 12697746
  • £64.65 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
8.5 in.
Holds # 20 to # 25 Blade
# 4
21.6 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, Holds No. 20-25 Blades > Holds No. 20-25 Blades