Learn More
Sheep anti-Rabbit IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP1-75353
150.40 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
IgG (H+L) | |
Polyclonal | |
HRP | |
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free | |
This Sheep anti-Rabbit IgG (H+L) Secondary Antibody [HRP] was developed against purified rabbit IgG (H&L). | |
1 mg | |
Adaptive Immunity, Immunology | |
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
LYOPH | |
Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin | |
Sheep | |
Affinity purified | |
RUO | |
Secondary | |
Rabbit | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.